Cole-Parmer Two-Channel Paperless Recording Systems
No more paper to jam, no more pens to clog
  • Intuitive operation, flexible input configurations, and large memory capabilities
  • Highly reflective LCD with bright white LED backlight displays real-time data in digital or trend format
  • Two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs, as well as frequency and pulse inputs
  • Four internal alarm set points, two alarm relay outputs, and one digital control input
  • Ethernet and USB communication options available
Compact size combined with extreme flexibility makes these two-channel paperless recorders a powerful, yet economical, digital data recording system. A maximum storage rate of 100 samples per second can be set for both channels, letting you capture short duration process signals anomalies. Use CompactFlash™ cards up to 2 GB for data storage over longer time periods.
$1,408.56CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Number of Channels Description
Availability Pricing
COMPARE
4350 2 Paperless Recorder, 2-Channel; Univ AC Adapter/Rechargeable Battery
$1,408.56CAD / Each