This product is now discontinued, View Related Products
Cole-Parmer® Essentials Tight-Ties Nylon Cable/Zip Ties
Chemical-resistant and corrosion-proof fasteners
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Tensile strength is 50 lbs
Use to fasten pressure, vacuum, and liquid tubing lines or bundle lines together. Use from -40 to 185°F (-40 to 85°C). When fastened, diameters adjust from 5/32" to 4" (2.6 to 10.2 cm).
$16.50 - $50.83CAD / Pkg of 400
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
COMPARE
14 Tight-Ties Nylon Cable/Zip Ties, Assortment; 400/Pk
$50.83CAD / Pkg of 400
COMPARE
4 Tight-Ties Nylon Cable/Zip Ties, 4\"; 100/Pk
$16.50CAD / Pkg of 100
COMPARE
5 5/8 Tight-Ties Nylon Cable/Zip Ties, 5.625\"; 100/Pk
$16.50CAD / Pkg of 100
COMPARE
8 Tight-Ties Nylon Cable/Zip Ties, 8\"; 100/Pk
$16.94CAD / Pkg of 100
COMPARE
11 Tight-Ties Nylon Cable/Zip Ties, 11\"; 50/Pk
$16.50CAD / Pkg of 50
COMPARE
14 Tight-Ties Nylon Cable/Zip Ties, 14\"; 50/Pk
$16.58CAD / Pkg of 50