Cole-Parmer® SHP-300 Series Digital Round-Top Stirring Hot Plates
Simple and precise stirring and heating at your fingertips
  • Innovative fingertip control even while wearing gloves
  • Advanced brushless motor provides quiet and worry-free operation
  • Sealed exterior keeps spills away for easy maintenance
  • LCD shows set and working parameters
  • Hot top warning lights protects you from accidental burns
  • Ceramic-coated aluminum top plate provides excellent heat transmission
Sleek, low-profile digital stirring hot plate features a unique control pad that allows all operations to be set using just your fingertip. An LCD shows both set and real-time parameters which are lockable to prevent accidental changes during use.
Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built-in timer can be set for up to 9 hours or continuous operation. Unit offers stirring speeds up to 1800 rpm with max capacity of 20 liters.
Optimal heating is produced by an integrated temperature sensor and dual PID control that rapidly heats plate and maintains desired temperature. Dual temperature control circuits also ensure temperature stability in case one circuit malfunctions.
Safety features include user-set safety temperature limit through main control panel, hot warning light indicator, and digital error code functions for easy troubleshooting.
The ceramic-coated aluminum plate offers excellent heat transmission, rapid heating with a 600-watt element, even temperature distribution, plus is chemical resistant and easy to clean.
$1,202.29CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Temperature (° F) Description
Availability Pricing
COMPARE
1800 644 Stirring Hotplate with Timer, 20L Capacity, 110V
$1,202.29CAD / Each
COMPARE
1800 644 Stirring Hotplate with Timer, 20L Capacity, 220V
$1,202.29CAD / Each