This product is now discontinued, View Related Products
Foxx Life Sciences Sterile Plastic Media Bottles
Safe alternative to glass
  • Use right out of the package—bottles come presterilized
  • Store tissue culture media, serum, and buffers
  • Manufactured in ISO Class 8 cleanroom
  • Comply with USP Class VI standards and are BPA-free
These plastic media bottles make storing your common life science solutions simple. Gamma sterilized for convenient use—right out of the package. Molded indents and graduations make handling and reading liquid levels easy. Innovative cap design allows for easy gripping, uncapping, and stacking.
Polycarbonate (PC) bottles are autoclavable and offer good alcohol resistance. Polystyrene (PS) bottles are ideal for aqueous cell culture solutions. Gas-impermeable PETG bottles offer low-temperature stability and handle your general lab solution storage needs.
$107.56 - $284.00CAD / Case of 24
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Autoclavable Capacity (mL) Material
Availability Pricing
COMPARE
Yes 250 Polystyrene
$107.56CAD / Case of 24
COMPARE
Yes 500 Polystyrene
$157.65CAD / Case of 24
COMPARE
Yes 1000 Polystyrene
$194.49CAD / Case of 24
COMPARE
No 250 Polyethylene Terephthalate Glycol
$138.50CAD / Case of 24
Foxx Life Sciences Media Bottle, PETG, 500 mL, Sterile; 24/CS RK-06060-90 Mfr # 1177-RLS
COMPARE
No 500 Polyethylene Terephthalate Glycol
$222.48CAD / Case of 24
Foxx Life Sciences Media Bottle, PETG, 1000 mL, Sterile; 24/CS RK-06060-91 Mfr # 1178-RLS
COMPARE
No 1000 Polyethylene Terephthalate Glycol
$284.00CAD / Case of 24
COMPARE
No 250 Polycarbonate(PC)
$141.45CAD / Case of 24
COMPARE
No 500 Polycarbonate(PC)
$200.38CAD / Case of 24
COMPARE
No 1000 Polycarbonate(PC)
$265.21CAD / Case of 24
Showing 1- 9 of 9