Cole-Parmer® Stackable Lab Sampling Scoops, Polypropylene
Stackable lab scoops save space and store easily
  • Highly resistant to solvents and chemicals
  • Withstand temperatures between -20 to 121 °C (-4 to 249.8 °F)
  • Sharp, flat edge helps obtain samples
Ideal for sampling various powder, granulate, and paste form samples, these polypropylene lab scoops resist most acids, bases, and many commonly used solvents. Scoops of the same size are easily stackable requiring less storage space, and they are guaranteed to withstand temperatures between -20 to 121 °C (-4 to 249.8 °F). The sharp edge of these laboratory scoops allows you to pick up all types of samples even from wet surfaces. An easy-to-grip handle provides comfortable use, and a flat bottom allows these lab scoops to stand horizontally on a flat surface and double as a weighing pan. These sampling scoops also have a smooth, easy to clean surface and can be autoclaved if needed for sterility.
When used in place of disposable scoops, these reusable lab scoops help reduce waste in the laboratory, as well as save you money and reordering time. Specified volumes are printed on the handles but are approximate and not intended for measuring specific volumes.
$13.41 - $80.89CAD / Pkg of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
2 6.2 2
$13.41CAD / Pkg of 12
COMPARE
5 8.2 2.6
$15.18CAD / Pkg of 12
COMPARE
10 9.7 3.4
$15.18CAD / Pkg of 12
COMPARE
25 13 4.5
$17.53CAD / Pkg of 12
COMPARE
50 16.5 5.3
$32.71CAD / Pkg of 12
COMPARE
100 20.2 7
$36.83CAD / Pkg of 12
COMPARE
250 26.2 9.1
$58.49CAD / Pkg of 12
COMPARE
500 31.6 15.5
$80.89CAD / Pkg of 12
COMPARE
1000 38.5 14.6
$57.46CAD / Pkg of 6
Showing 1- 9 of 9