This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heaters (Analog)
Precision temperature stability ensures reproducibility of results
  • Interchangeable modular heating blocks provide flexibility for heating different-size tubes -order blocks separately
Designed for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures. Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Create a versatile system unique to your application; order either an analog or digital heater and the required heating block module (sold separately online).
CAUTION! To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed to run as a dry bath / incubator.
Analog Dry Block Heaters are economical, high-wattage, constant temperature heaters that are ideal for a wide variety of laboratory applications. They are controlled by two separate temperature adjustment knobs, with 1 to 10 dial markings for low and high temperature ranges. Low-range knob adjusts from ambient to 100ºC and high-range knob adjusts from 75 to 150ºC. A separate thermometer or probe is needed to set and verify the setpoint temperature.
$412.55 - $1,583.89CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Number Of Blocks Power (VAC)
Availability Pricing
COMPARE
1 120
$412.55CAD / Each
COMPARE
1 230
$412.55CAD / Each
COMPARE
2 120
$636.50CAD / Each
COMPARE
2 230
$636.50CAD / Each
COMPARE
3 120
$816.26CAD / Each
COMPARE
3 230
$816.26CAD / Each
COMPARE
4 120
$1,026.95CAD / Each
COMPARE
4 230
$1,026.95CAD / Each
COMPARE
6 120
$1,583.89CAD / Each
COMPARE
6 230
$1,583.89CAD / Each
Showing 1- 10 of 10