This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Heating Blocks for Microcentrifuge Tubes
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Microcentrifuge tube blocks are single size blocks and are designed to hold a variety of tapered microcentrifuge tubes. Blocks are available to hold sizes from 0.5mL to 2.0mL.
$162.07CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mL) Description
Availability Pricing
COMPARE
30 0.5 Single Block for Microcentrifuge Tubes, 0.5 ml Tube
$162.07CAD / Each
COMPARE
20 1.5 Single Block for Microcentrifuge Tubes, 1.5 ml Tube
$162.07CAD / Each
COMPARE
20 1.5 Heat Block; 1.5 ml EppenDOrf Microcentrifuge Tubes
$162.07CAD / Each
COMPARE
20 2 Heat Block; 2.0 ml EppenDOrf Microcentrifuge Tubes
$162.07CAD / Each
COMPARE
20 2 Heat Block; 2.0 ml Microcentrifuge Tubes
$162.07CAD / Each