This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Heating Blocks for Centrifuge Tubes
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Centrifuge Tube blocks are single size blocks and are available in 15mL or 50mL tube sizes.
$162.07CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mL)
Availability Pricing
COMPARE
12 15
$162.07CAD / Each
COMPARE
5 50
$162.07CAD / Each