Cole-Parmer® Spex® BM-600 Planetary Mill Accessories
Accessories to rapidly reduce hard and brittle samples with the BM-600 Planetary Mill
  • Jar rotation in combination with grinding balls impacting the sample and the walls of the jar allow for both milling and mixing
  • Excellent for grinding rocks, minerals, sand, cement, slag, ceramics, glass, and pharmaceuticals
These accessories are compatible with the Cole-Parmer® BM-600 Planetary Mills 04900-16 and 04900-17. Mill sold separately.
Choose from 250 mL or 500 mL jar capacities and stainless steel or hardened steel jars and grinding media. Generally, the grinding jar and grinding balls are made from the same material.
$77.35 - $3,867.65CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
BM-600 Planetary Ball Mill Grinding Jar, Hardened Steel, 250 mL
$2,320.59CAD / Each
COMPARE
BM-600 Planetary Ball Mill Grinding Jar, Hardened Steel, 500 mL
$2,784.71CAD / Each
COMPARE
BM-600 Planetary Ball Mill Grinding Jar, Stainless Steel, 250 mL
$3,094.12CAD / Each
COMPARE
BM-600 Planetary Ball Mill Grinding Jar, Stainless Steel, 500 mL
$3,867.65CAD / Each
COMPARE
250 mL Grinding Jar O-rings for 250 mL Grinding Jar; 10/Pk
$77.35CAD / Pkg of 10
COMPARE
500 mL Grinding Jar O-rings for 500 mL Grinding Jar; 10/Pk
$108.29CAD / Pkg of 10
COMPARE
500 mL Grinding Jar Hardened Steel Ball Set, 40mm; 4/Pk
$232.06CAD / Pkg of 4
COMPARE
250 and 500 mL Grinding Jar Hardened Steel Ball Set, 30mm; 8/Pk
$216.59CAD / Pkg of 8
COMPARE
250 and 500 mL Grinding Jar Hardened Steel Ball Set, 20mm; 20/Pk
$208.85CAD / Pkg of 20
COMPARE
250 and 500 mL Grinding Jar Hardened Steel Ball Set, 10mm; 100/Pk
$208.85CAD / Pkg of 100
COMPARE
500 mL Grinding Jar Stainless Steel Ball Set, 40mm; 4/Pk
$402.24CAD / Pkg of 4
COMPARE
250 and 500 mL Grinding Jar Stainless Steel Ball Set, 30mm; 8/Pk
$324.88CAD / Pkg of 8
COMPARE
250 and 500 mL Grinding Jar Stainless Steel Ball Set, 20mm; 20/Pk
$293.94CAD / Pkg of 20
COMPARE
250 and 500 mL Grinding Jar Stainless Steel Ball Set, 10mm; 100/Pk
$324.88CAD / Pkg of 100
COMPARE
250 mL Grinding Jar Jar Sieve for 250 ml Jar
$1,082.94CAD / Each
COMPARE
500 mL Grinding Jar Jar Sieve for 500 ml Jar
$1,160.29CAD / Each
Showing 1- 16 of 16