This product is now discontinued, View Related Products
Cole-Parmer® Single-Channel Adjustable-Volume Pipettors
Excellent accuracy and performance at an economical price
  • Autoclavable tip cone and lower section assembly
  • Tip ejector with blow-out step
  • Individually calibrated in accordance with DIN 12650
Ergonomic, lightweight design with soft plunger movement minimizes hand fatigue. Pipets feature streamlined, low-pressure tip ejector for easy access to narrow containers. They facilitate last drop dispensing and reverse pipetting technique with two-stage plunger operation for blow-out. Easy to calibrate and disassemble for routine servicing or autoclaving. Adjustable-volume pipettors feature quick, easy click-stop volume settings.
$269.93 - $271.10CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Autoclavable Max Volume (mL) Min Volume (mL)
Availability Pricing
COMPARE
Yes 0.01 0.0001
$269.93CAD / Each
COMPARE
Yes 0.05 0.005
$271.10CAD / Each
COMPARE
Yes 0.1 0.01
$271.10CAD / Each
COMPARE
Yes 0.2 0.02
$271.10CAD / Each
COMPARE
Yes 1 0.1
$271.10CAD / Each
COMPARE
Yes 5 0.5
$271.10CAD / Each