Cole-Parmer® MSHM-200 Series Magnetic Stirring/Heating Mantles
Magnetic stirrer and heating mantle in one convenient unit
  • Flexible coiled heating element wraps the heater around round-bottom flask to optimize heat transfer
  • “Cool to the touch” exterior provides safe handling
These stirring/heating mantles combine the functionality of a heating mantle with a magnetic stirrer providing even heating and mixing. Element temperatures can reach up to 842°F (450°C) yet the exterior remains cool to the touch. For maximum safety, the flask does not come in direct contact with the heating element. Built-in energy regulator and safety earth (ground) screen offer added safety protection.
Reach stirring speeds up to 2000 rpm. Simply use the auto-recapture switch to reactivate stirring if stirring is interrupted and stirring motion is lost. Two colored lights by each control dial indicate "power on" and "heat on." Powder-coated aluminum housing makes it easy to clean. All heating mantles feature a support bracket for holding a support rod.
$1,337.84 - $2,386.89CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Max temperature (° F) Power (VAC)
Availability Pricing
COMPARE
100 842 230
$1,395.30CAD / Each
COMPARE
100 842 115
$1,337.84CAD / Each
COMPARE
250 842 230
$1,709.13CAD / Each
COMPARE
250 842 115
$1,709.13CAD / Each
COMPARE
500 842 230
$1,463.08CAD / Each
COMPARE
500 842 115
$1,438.03CAD / Each
COMPARE
1000 842 230
$1,449.82CAD / Each
COMPARE
1000 842 115
$1,431.40CAD / Each
COMPARE
5000 842 230
$2,386.89CAD / Each
COMPARE
5000 842 115
$2,386.89CAD / Each
Showing 1- 10 of 10