This product is now discontinued, View Related Products
Cole-Parmer Rugged Digital Pressure Gauges
±0.25% test gauge accuracy
  • 316 stainless steel wetted parts
  • Fast response—3 readings per second
  • Temperature-compensated piezoresistive sensor
  • Factory-calibrated using NIST traceable standards
Note: NIST calibration report is NOT provided - order separately below
$518.63 - $725.64CAD / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.25% full-scale 300 0.1 psig
$719.01CAD / Each
COMPARE
±0.25% full-scale 100 0.1 psig
$704.72CAD / Each
COMPARE
±0.25% full-scale 500 0.1 psig
$711.21CAD / Each
COMPARE
±0.25% full-scale 1000 1 psig
$725.64CAD / Each
COMPARE
±0.25% full-scale 10000 1 psig
$720.49CAD / Each
COMPARE
±0.25% full-scale 0.01 kg/cm²
$720.49CAD / Each
COMPARE
±0.25% full-scale ±1 digit 3 0.01 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 5 0.01 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 15 0.1\" Hg, 0.1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 15 0.01 psi
$521.29CAD / Each
COMPARE
±0.25% full-scale ±1 digit 30 0.1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 100 0.1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 199.9 0.1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 300 1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 500 1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 1000 1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 3000 1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 5000 1 psi
$518.63CAD / Each
COMPARE
±0.25% full-scale ±1 digit 1 Torr
$604.09CAD / Each
COMPARE
±0.25% full-scale ±1 digit 0.1\" Hg
$604.09CAD / Each
Showing 1 - 20 of 50