Cole-Parmer® RS-250 Series Reaction Stations
Run multiple reactions in a single station, saving you time and money
  • Excellent temperature uniformity across the block
  • Soft-start feature minimizes splashing
Run these reaction stations at the same temperature and stirring rates at several positions simultaneously. They are great for a wide range of applications from simple synthesis to process optimization. High-performance magnetic stirrers ensure maximum coupling between the stir bar in the sample and motor.
Easily set and view temperature and stir rate on the digital display. Use the RS-232 or RS-485 connections to interface with a robotic workstation or a computer. The well-insulated stations stay cool to the touch for safe handling. PTFE coating on vessel block provides protection from chemical spills.
$8,855.07 - $21,894.58CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Combination Block -Tube / Vial Size Accepted Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
1 to 25 mL 150 230
$8,855.07CAD / Each
COMPARE
Interchangeable blocks (order separately) 150 115
$21,894.58CAD / Each