This product is now discontinued, View Related Products
Cole-Parmer Roto-Torque Rotator 47874
Gentle rotating motion efficiently mixes the contents of your test tubes, flasks and bottles
  • Lock at the desired angle with the adjustable tilt stand (included)
Shakers have a speed range from 3 to 80 rpm. Rotators require one of the three attachments (sold separately) to hold your samples.
What's Included: model 07637-21 includes a 6-ft, three-wire cord with plug; model 07637-22 has a stripped end cord.
$14.72 - $1,281.85CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Roto-Torque Variable Speed Rotator, 115 VAC, 50/60 Hz 07637-21
$14.72CAD / Each
COMPARE
Roto-Torque Variable Speed Rotator, 230 VAC, 50/60 Hz
$1,281.85CAD / Each
COMPARE
Multipurpose Disc for Roto-Torque Rotator
$643.87CAD / Each
COMPARE
Clips for Roto-Torque Rotator, fit 10 to 15 mm tubes
$151.02CAD / Dozen of 1
COMPARE
Clips for Roto-Torque Rotator, fit 15 to 20 mm tubes
$151.02CAD / Dozen of 1
COMPARE
Tube Clips 20-25mm Size
$36.83CAD / Dozen
COMPARE
Tube Clips 25-40mm Size
$44.20CAD / Dozen
Showing 1- 7 of 7