This product is now discontinued, View Related Products
Cole-Parmer Rocking Shakers
Slow speeds mix live cultures without agitation
  • Platform lip and nonskid mat securely hold flat-bottom vessels
  • Built-in timer and digital speed display ensure repeatable results
  • Triple the capacity with optional tier system
These rocking shakers offer small capacity options for personal use or for tight spaces, and larger capacities for larger laboratories and high-throughput applications. Easily disassemble and clean the tier system, which is held together by magnets (order separately).
See-saw motion rocker (51700-21 to -24) is perfect for culture flasks and Petri dishes, producing a rolling wave in your sample.
Gyrational motion rocker (51700-17 to -20) is ideal for low-foam applications requiring gentle swirling, such as DNA extractions and histological preparations.
$1,081.47 - $2,070.11CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Load (lbs) Power (VAC)
Availability Pricing
COMPARE
6.6 120
$1,081.47CAD / Each
COMPARE
6.6 230
$1,547.06CAD / Each
COMPARE
22 120
$2,070.11CAD / Each
COMPARE
22 230
$2,033.28CAD / Each