This product is now discontinued, View Related Products
Cole-Parmer Reciprocating Shakers
Quiet enough for continuous operation
  • Microprocessor-controlled speed maintains consistent shaking
  • Soft-start function brings motor up to speed gradually, preventing sample loss
  • Built-in digital timer and continuous operation mode adapt to any mixing protocol
  • Fully adjustable vessel containment system fits most flasks and bottles
These reciprocating shakers feature adjustable speed with fixed stroke length, mixing gently to vigorously. Horizontal and vertical adjustments to the platform securing bars accommodate nearly any flask or bottle. Holds up to twelve 250-mL, nine 500-mL, four 1-L, or two 2-L vessels.
Digital timer sets from 1 to 999 minutes; when the counter reaches zero the shaker comes to a stop, then sounds an alert. Microprocessor maintains speed accuracy over long runs including continuous operation.
$2,092.21 - $2,423.73CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
$2,423.73CAD / Each
COMPARE
230
$2,092.21CAD / Each