This product is now discontinued, View Related Products
Cole-Parmer Polystat Stainless Steel Heated Circulating Baths
Quality performance at an affordable price!
  • Easy-to-use PID controller with moisture-tight LCD (0.01°) in °C, °F, or K
  • Two-speed pump includes plumbing connections for external circulation
  • Safety features include high-temperature cutoff and acoustic alarms
These baths are ideal for applications with a wide variety of temperature control requirements up to 200°C. Available with tank capacity up to 6.5, 9.5 or 19.6 liters, the compact footprint maximizes bench space. Bright digital display allows for simple operation. Three languages: English, French, and German.
Industrial-grade circulating pumps deliver extra flow and pressure providing more efficient heat exchange, better temperature uniformity, and improved reservoir agitation. Long-life, high wattage heaters for applications requiring rapid temperature recovery from fluctuations.
$2,890.79 - $6,894.73CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
150 115 6.5
$3,362.28CAD / Each
COMPARE
150 230 6.5
$3,539.08CAD / Each
COMPARE
200 115 6.5
$4,399.54CAD / Each
COMPARE
200 230 6.5
$6,247.91CAD / Each
COMPARE
150 115 9.5
$3,771.14CAD / Each
COMPARE
150 230 9.5
$5,174.55CAD / Each
COMPARE
200 115 9.5
$5,176.76CAD / Each
COMPARE
200 230 9.5
$6,541.85CAD / Each
COMPARE
150 115 19.6
$4,277.99CAD / Each
COMPARE
150 230 19.6
$5,690.97CAD / Each
COMPARE
200 115 19.6
$5,013.95CAD / Each
COMPARE
200 230 19.6
$6,894.73CAD / Each
COMPARE
100 115 6.5
$2,890.79CAD / Each
COMPARE
100 230 6.5
$3,327.65CAD / Each
COMPARE
100 115 9.5
$3,785.14CAD / Each
COMPARE
100 230 9.5
$5,970.91CAD / Each
COMPARE
100 115 19.6
$3,889.75CAD / Each
COMPARE
100 230 19.6
$4,234.52CAD / Each
Showing 1- 18 of 18