This product is now discontinued, View Related Products
Cole-Parmer Polystat PPO Open Heating Circulating Baths
Economical alternative to stainless steel baths for less demanding applications
  • Three programmable set points
  • Two-speed pump with adjustable nozzle and up to 15 L/min flow
  • Plumbed inlet/outlet connections for external circulation
These versatile baths provide exceptional features and flexibility. Bright digital display allows for simple operation and more precise control. High-temperature cutoff protects your samples and equipment from damage. Rugged PPO baths allow for heating to 100°C and are the most economical choice.
$2,326.48 - $2,959.30CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
60 115 5.3
$2,414.15CAD / Each
COMPARE
50 230 5.3
$2,326.48CAD / Each
COMPARE
60 115 14.1
$2,862.06CAD / Each
COMPARE
50 230 14.1
$2,778.81CAD / Each
COMPARE
60 115 21
$2,959.30CAD / Each
COMPARE
50 230 21
$2,873.11CAD / Each