This product is now discontinued, View Related Products
Cole-Parmer Polystat Heating/Cooling Bath Starter Kits
Convenient bundled kits to get you started!
  • Powerful refrigeration system provides excellent cooling capacity across range of operating temperatures
  • Two-speed pump with up to 21 L/min flow
  • User-friendly interface available in three languages: English, French, German
  • Compact footprint maximizes bench space
Bright digital display and intuitive interface allow for simple operation. These recirculating bath kits are ideal for industrial or laboratory applications with a wide variety of temperature control requirements. Choose from a standard 15-L kit, or a premium 6-L kit with PC monitoring and mobility from the included trolley.
The included bath fluids cover a wide range of temperatures, including the range of the baths. SIL 180 fluid can handle temperatures from -40 to 200°C, and the ethylene glycol mixture can handle -30 to 90°C. Plumbing kits come insultated for use at high or low temperatures.
$6,424.72 - $9,734.69CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
200 -28 Standard Starter Kit, 15-L Heat/Cool Bath, -28 to 200C; 115VAC
$9,734.69CAD / Each
COMPARE
100 -20 Premium Starter Kit, 6-L Heat/Cool Bath, -20 to 100C; 115VAC
$6,424.72CAD / Each