Cole-Parmer® CBH-400 Series Open Heated Circulating Bath Systems
Full-range cooling for quick ramp up and cool-down – saves valuable time
  • Robust cooling and heating from -10 to 80°C
  • Compact design maximizes bench space and provides quick temperature changes
  • Internal tank design minimizes fluid evaporation
True thermostatic control laboratory chiller. Removes or adds temperature as required by your application. This closed laboratory chiller is ideal for low load applications such as cooling smaller analytical instrumentation, jacketed reactors, and rotary evaporators. This system can also be used as an economical replacement to tap water cooling in small laboratory applications. Rapid cool-down and heat-up times enable you to maintain temperature to ±0.1°C. Powerful force/suction pump provides flow rates up to 21 L/min at pressures up to 11.7 psi (805 mbar). All models have USB ports for applications that require serial communications.
$6,939.67 - $7,190.14CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Description
Availability Pricing
COMPARE
250 115 recirculator, 17 L/min, 250W cooling capacity, 115V 60 Hz
$6,976.50CAD / Each
COMPARE
250 230 recirculator, 17 L/min, 250W cooling capacity, 230V 50 Hz
$6,976.50CAD / Each
COMPARE
250 115 Recirculator w/ Force/Suction Pump, 250W; 115V
$6,939.67CAD / Each
COMPARE
250 230 Recirculator w/ Force/Suction Pump, 250W; 230V
$6,939.67CAD / Each
COMPARE
500 115 recirculator, 17 L/min, 500W cooling capacity, 115V 60 Hz
$6,954.40CAD / Each
COMPARE
500 115 Recirculator w/ Force/Suction Pump, 500W; 115V
$7,190.14CAD / Each
COMPARE
500 230 Recirculator w/ Force/Suction Pump, 500W; 230V
$7,168.04CAD / Each
Showing 1- 7 of 7