This product is now discontinued, View Related Products
Cole-Parmer PolyStat Acrylic Open Heating Circulating Baths
Economical alternative to stainless steel baths for less demanding applications
  • Three programmable set points
  • Two-speed pump with adjustable nozzle and up to 17 L/min flow
  • Plumbed inlet/outlet connections for external circulation
These versatile baths provide exceptional features and flexibility. Bright digital display and intuitive controller allow for simple operation and more precise control. High-temperature cutoff protects your samples and equipment from damage. Transparent acrylic baths handle temperatures to 80°C and provide visibility of your bathing samples.
6 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
60 115 7
$2,655.79CAD / Each
COMPARE
50 230 7
$3,591.39CAD / Each
COMPARE
60 115 12.8
$2,893.00CAD / Each
COMPARE
50 230 12.8
$3,865.44CAD / Each
COMPARE
60 115 19
$3,942.05CAD / Each
COMPARE
50 230 19
$2,962.99CAD / Each