This product is now discontinued, View Related Products
Cole-Parmer Plexiglas Racks
Interchangeable racks hold a variety of tubes and vials
Plexiglas Racks for 6- and 8-L Tanks are available with different hole diameters to hold test tubes, scintillation vials, or 1.5-mL microcentrifuge tubes. Racks attach to the tank sides and suspend tubes or vials in bath. The 6-L tank holds up to three racks without a circulator or two racks with a circulator. The 8-L tank holds up to four racks without a circulator or three racks with a circulator. Circulator sold separately.
$123.76 - $175.33CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Plexiglas Racks for 6 Tubes with 30 mm diameter
$123.76CAD / Each
COMPARE
Plexiglas Racks for 12 Tubes with 23 mm diameter
$169.44CAD / Each
COMPARE
Plexiglas Racks for 20 Tubes with 17 mm diameter
$175.33CAD / Each
COMPARE
Plexiglas Racks for 20 Tubes with 18 mm diameter
$173.86CAD / Each
COMPARE
Plexiglas Racks for 30 Tubes with 13 mm diameter
$173.86CAD / Each
COMPARE
Plexiglas Racks for 36 Tubes with 11 mm diameter
$169.44CAD / Each