Cole-Parmer Pipette Tips
Cleanroom produced for ready-to-use applications
  • Constructed of pure, high-quality polypropylene without additives
  • Slim, thin-walled tips feature graduations
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
Designed to give high levels of accuracy, precision, and reproducibility, these polypropylene (PP) tips are suitable for use with most pipettors. The smooth surface minimizes binding of enzymes and nucleic acids to the walls.
Tips are available packaged in a bulk bag or in a storage box with 96-tip rack insert and hinged lid. Order environmentally friendly refill racks separately.
$39.04 - $182.70CAD / Pkg of 1000
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Package Of Sterile
Availability Pricing
COMPARE
5000 Bag of 200 No
$182.70CAD / Pkg of 200
COMPARE
20 Bag of 1000 No
$106.08CAD / Pkg of 1000
COMPARE
20 Bag of 1000 No
$68.51CAD / Pkg of 1000
COMPARE
50 Bag of 1000 No
$53.41CAD / Pkg of 1000
COMPARE
200 Bag of 1000 No
$50.46CAD / Pkg of 1000
COMPARE
300 Bag of 1000 No
$50.46CAD / Pkg of 1000
COMPARE
1000 Bag of 1000 No
$39.04CAD / Pkg of 1000
COMPARE
20 5 racks of 96 each No
$94.30CAD / Pkg of 480
COMPARE
20 5 racks of 96 each No
$99.45CAD / Pkg of 480
COMPARE
50 5 racks of 96 each No
$94.30CAD / Pkg of 480
COMPARE
200 5 racks of 96 each No
$94.30CAD / Pkg of 480
COMPARE
300 5 racks of 96 each No
$79.56CAD / Pkg of 480
Showing 1- 12 of 12