This product is now discontinued, View Related Products
Cole-Parmer Pipette Filter Tip Refill Racks
Reduce waste by refilling
  • Cleanroom produced for ready-to-use applications
  • Slim, thin-walled tips feature graduations
  • Filter protects samples and pipettor against contamination
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
Constructed of pure, high-quality polypropylene (PP) without additives, these tips are designed to give high levels of accuracy, precision, and reproducibility. The smooth surface minimizes binding of enzymes and nucleic acids to the tip wall.
The non-self-sealing filter plug prevents aerosols and liquids from being aspirated into the body of the pipettor. Filter is free from chemical additives, does not swell, and is made of pure polyethylene (PE).
Each 96-tip refill rack comes in an environmentally friendly package—simply insert rack into an existing storage box.
$135.55 - $282.89CAD / Pkg of 960
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Min Volume (µL) Sterile
Availability Pricing
COMPARE
1 0.1 No
$221.01CAD / Pkg of 960
COMPARE
10 0.5 No
$235.74CAD / Pkg of 960
COMPARE
20 1 No
$166.49CAD / Pkg of 960
COMPARE
20 2 No
$166.49CAD / Pkg of 960
COMPARE
100 5 No
$135.55CAD / Pkg of 960
COMPARE
200 5 No
$151.76CAD / Pkg of 960
COMPARE
1000 50 No
$172.83CAD / Pkg of 960
COMPARE
1 0.1 Yes
$244.14CAD / Pkg of 960
COMPARE
10 0.5 Yes
$282.89CAD / Pkg of 960
COMPARE
20 1 Yes
$206.27CAD / Pkg of 960
COMPARE
20 Yes
$206.27CAD / Pkg of 960
COMPARE
100 5 Yes
$185.65CAD / Pkg of 960
COMPARE
200 5 Yes
$185.65CAD / Pkg of 960
COMPARE
1000 50 Yes
$185.65CAD / Pkg of 960
Showing 1- 14 of 14