This product is now discontinued, View Related Products
Cole-Parmer Penny Head Hollow Stoppers, PTFE
Snug fit and easy removal – will not freeze or bind
Durable, chemically inert white PTFE stoppers are ideal for use with flasks, funnels, and cylinders that require flask-length stoppers. Hollow top minimizes expansion/contraction caused by heating or cooling. You can also use the hollow top as a container for micro-analytical processing of solutions.
$32.05 - $84.72CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Standard Size Number
Availability Pricing
COMPARE
9
$33.15CAD / Each
COMPARE
13
$32.05CAD / Each
COMPARE
16
$34.62CAD / Each
COMPARE
19
$40.52CAD / Each
COMPARE
22
$49.73CAD / Each
COMPARE
27
$72.93CAD / Each
COMPARE
32
$84.72CAD / Each
COMPARE
38
$51.57CAD / Each
Showing 1- 8 of 8