This product is now discontinued, View Related Products
Cole-Parmer Multichannel Controllers
Ideal for multiple level monitoring and control applications
These easy-to-use multichannel controllers accept 4-20 mA, MODBUS, and flowmeter pulse inputs and display them on a large backlit LCD screen. The main screen displays all the inputs in both numeric and bargraph format and its individual screens display detailed information about each input. These models are suitable for all general-purpose monitoring and controlling applications such as flow, level, pressure, temperature and vibration. The MODBUS RTU feature makes these units ideal for SCADA applications.
These controllers have excellent graphical display and interface with large bargraph display to quickly help spot trends or problems. There are screens which show Input State, Flow Meter Input Values, as well as a detailed numeric screens for each input, advanced operational features without all the programming required by a PLC.
The display is readable in direct sunlight and the on-screen set-up is easy to configure with intuitive menus and detailed screens. Programming and data logging PC-based software comes with the controller.
$455.28 - $2,591.69CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type Max DC Voltage VDC (VDC)
Availability Pricing
COMPARE
Pulse (100mVp-p to 15Vp-p)
$2,134.94CAD / Each
COMPARE
Pulse (100mVp-p to 15Vp-p)
$2,134.94CAD / Each
COMPARE
Pulse (100mVp-p to 15Vp-p)
$2,591.69CAD / Each
COMPARE
Pulse (100mVp-p to 15Vp-p)
$2,591.69CAD / Each
COMPARE
$455.28CAD / Each