Cole-Parmer® OVF-400 Series Mechanical Convection Drying Ovens
Quickly heat and dry samples
  • Stainless steel inner chamber resists corrosion
  • Rounded inner chamber corners offer easy cleaning
  • Able to heat samples up to 482°F (250°C)
Mechanical convection ovens feature a PID controller which provides you accurate and reliable temperature control up to 482°F (250°C). Ideal for drying and sterilization. The ovens require minimal heating time, reaching 212°F (100°C) in 20 to 50 minutes. Easy-to-view LCD reads time, current temperature, set temperature, running status, and fan status.
Silicone rubber gasket offers a tight seal, ensuring you good heat retention and low energy consumption. The gasket comes with a long service life and can be easily replaced when needed. Additional oven features include auto-start after power loss/return, temperature deviation alarm, overtemperature set point alarm, and overcurrent protection. An air vent located at the back of the unit can be adjusted by the knob located in front of the oven to control inner air and vapor circulation.
$1,197.13 - $7,655.73CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC) Temperature Uniformity
Availability Pricing
COMPARE
16 120 ±1.5% at 100°C
$1,197.13CAD / Each
COMPARE
16 220 ±1.5% at 100°C
$1,197.13CAD / Each
COMPARE
30 120 ±2% at 100°C
$1,403.40CAD / Each
COMPARE
30 220 ±2% at 100°C
$1,403.40CAD / Each
COMPARE
50 120 ±2.5% at 100°C
$1,654.62CAD / Each
COMPARE
50 220 ±2.5% at 100°C
$1,654.62CAD / Each
COMPARE
80 120 ±3% at 100°C
$1,860.89CAD / Each
COMPARE
136 120 ±3% at 100°C
$2,408.99CAD / Each
COMPARE
136 220 ±3% at 100°C
$2,410.47CAD / Each
COMPARE
220 220 ±3% at 100°C
$2,867.22CAD / Each
COMPARE
400 220 ±3.5% at 100°C
$6,824.74CAD / Each
COMPARE
620 220 ±4% at 100°C
$7,655.73CAD / Each
Showing 1- 12 of 12