Cole-Parmer® MB-800 Series Touch-Screen Moisture Balances
Four drying modes—standard, mild, fast, and step—accurately determine moisture content in various substances
  • Color capacitive touch-screen display provides clear visibility of weighing results
  • IR proximity sensors offer programmable touch-free balance control
  • USB, RS-232, Ethernet, and Wi-Fi interfaces offer secure connection with peripheral device
  • External calibration ensures accurate weighing
  • Weighing and drying working modes offer specific functions to meet individual needs
  • GLP function quickly diagnoses weighing errors
  • Numerous weighing units plus two custom user-defined units cover most measurement needs
  • Import and export records from 8 different databases via USB flash drive
  • Filter settings can be easily adjusted depending on the working environment
  • Menu screen can be customized to match your exclusive requirements
  • Displays and prints in 11 languages
The Cole-Parmer moisture analyzer balances are designed to precisely verify relative moisture and dry mass content in small samples of various substances, as well as determine the mass of weighed objects. TThe drying chamber features an IR Emitter which allows for quick, uniform sample drying and measurements; no mathematical calculations are required. The system allows you to select one of four drying profiles: standard, quick, mild, and step. Drying time is set by selecting either automatic, time-defined, manual, or user-defined finish mode.
Large 4-1/4” x 2-1/2” color capacitive touch-screen display makes operating the balances easy and can be customized to meet your personal programming needs. Two built-in infrared (IR) proximity sensors located on each side of the menu display can be programmed with different functions for each working mode, enabling touch-free balance control while minimizing cross-contamination and wear and tear on the buttons. The balances feature two working modes: weighing and drying. To achieve the best possible weighing conditions, a filter level setting can be adjusted to fast or slow mode depending on the working environment. Other features include quick-access keys, balance taring, balance zeroing, auto switch-off, and backlight turn-off time.
The balance offers several databases for recording products, users, packaging, customers, drying modes, drying reports, ambient conditions, and weighing. Up to 200 drying modes can be recorded, configured, and used. Only users with appropriate access can add new records, export data, import data, delete records, and print saved data. A RS-232, two USB interfaces, Ethernet port, and Wi-Fi offer external connection to a computer or printer for GLP/GMP-compliant output. The autotest GLP function provides an enhanced balance summary to aid in assessing operation and diagnosing reasons for occurrence of errors in weighing.
$4,903.44 - $5,373.45CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Heater Type Readability (g)
Availability Pricing
COMPARE
50 Infrared 0.0001
$5,373.45CAD / Each
COMPARE
50 Infrared 0.001
$4,903.44CAD / Each
COMPARE
210 Infrared 0.001
$4,919.65CAD / Each