Cole-Parmer® LB-400 Series Precision Toploading Balances
Quality and precision at an economical price
  • 5-year industry-leading warranty
  • Large, backlit LCD provides clear visibility of weighing results
  • Internal and external calibration options
  • Removable draft shield on milligram (0.001g) readability options
  • USB, RS-232, Ethernet, and Wi-Fi† interfaces
  • Filter settings can be easily adjusted depending on the working environment
  • Multiple working modes handle a variety of weighing functions
  • Fast stabilization and response time
The Cole-Parmer LT series of precision toploading balances provide advanced weighing features at an economical price. The large, stainless steel weigh pan allows for easy weighing of large items. Easy to operate, the balances are calibrated either internally or externally using a calibration mass. The large, backlit LCD allows a clear presentation of the weighing results.
To achieve the best possible weighing conditions, the balances feature a filter level setting that can be adjusted to a fast or slow mode depending on the working environment. Other features include quick access keys, balance taring, balance zeroing, auto switch-off, and backlight turn-off time.
The balance features several databases for recording users, products, weighings, and tares. Only users with appropriate access can adjust balance settings, export data, import data, and delete records. Two USB, two RS-232 interfaces, Ethernet, and Wi-Fi† port offer external connection to a computer or printer for GLP/GMP-compliant output.
†Only models with internal calibration feature Wi-Fi interface.
$2,230.71 - $3,158.95CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
COMPARE
Internal 360 Precision Toploading Balance with LCD, 360g x 1mg, Internal Calibration
$2,809.75CAD / Each
COMPARE
Internal 600 Precision Toploading Balance with LCD, 600g x 1mg, Internal Calibration
$2,962.99CAD / Each
COMPARE
Internal 2100 Precision Toploading Balance, 2100g x 0.01g, Internal Calibration
$2,652.10CAD / Each
COMPARE
Internal 4500 Precision Toploading Balance, 4500g x 0.01g, Internal Calibration
$2,982.14CAD / Each
COMPARE
Internal 6000 Precision Toploading Balance, 6000g x 0.01g, Internal Calibration
$3,158.95CAD / Each
COMPARE
External 360 Precision Toploading Balance, 360g x 1mg, External Calibration
$2,246.92CAD / Each
COMPARE
External 600 Precision Toploading Balance, 600g x 1mg, External Calibration
$2,392.79CAD / Each
COMPARE
External 2100 Precision Toploading Balance, 2100g x 0.01g, External Calibration
$2,230.71CAD / Each
COMPARE
External 4500 Precision Toploading Balance, 4500g x 0.01g, External Calibration
$2,520.97CAD / Each
COMPARE
External 6000 Precision Toploading Balance, 6000g x 0.01g, External Calibration
$2,871.64CAD / Each
Showing 1- 10 of 10