This product is now discontinued, View Related Products
Cole-Parmer® CH-400 Series Low-Temperature Benchtop Chillers
Reliable, precise cooling control in a space saving package
  • Easy to use interface
  • Quiet, energy efficient operation
  • Illuminated fluid-level indicator for easy viewing
  • Capable of cooling two rotary evaporators concurrently
These compact yet powerful chillers are designed for daily operation, with a simple interface and easy to read display for quick setup. Built-in security/safety with adjustable temperatures limits to warn if temperature rises to high or low, and local lockout to prevent unauthorized changes.
Features a 0.7 gal (2.65 L) reservoir with illuminated level indicator and a top-mounted fill port with built-in fluid filter. Easy-access reusable air filter keeps chiller running at peak efficiency. PID temperature controller offers stability of ±0.1°C.
Ideal for use with rotary evaporators, vacuum systems, and analytical equipment.
$4,408.38 - $8,691.53CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (L/min) Min Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
7.9 -10 120
$6,653.83CAD / Each
COMPARE
13.2 -10 120
$7,349.27CAD / Each
COMPARE
6.8 -10 240
$6,653.83CAD / Each
COMPARE
11.4 -10 240
$7,349.27CAD / Each
COMPARE
13.2 –20 120
$8,144.90CAD / Each
COMPARE
9.8 –20 120
$8,691.53CAD / Each
COMPARE
11.7 –20 240
$8,144.90CAD / Each
COMPARE
8.3 –20 240
$8,691.53CAD / Each
COMPARE
7.9 -5 120
$4,408.38CAD / Each
COMPARE
13.2 -5 120
$4,860.71CAD / Each
COMPARE
6.8 -5 240
$4,408.38CAD / Each
COMPARE
11.4 -5 240
$4,860.71CAD / Each
Showing 1- 12 of 12