This product is now discontinued, View Related Products
Cole-Parmer Low Level Sodium ISE
Accurately monitor sodium levels in the ppb range
  • Ideal for power plant monitoring of steam turbines and stainless steel generators
  • Sodium ion range: 0.1 ppb to 10 ppm
Economical alternative to accurately detect sodium ion impurities in the ppb range. Early detection helps to prevent sodium ion stress, corrosion, and cracking. Glass body combination electrode has an ammonia chloride reference solution. Rlectrode is avaliable with a BNC connector or a S7/detachable lead connection.
$889.93 - $920.87CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Dissolved Oxygen (mg/L (ppm)) Max pH (pH) Output Current
Availability Pricing
COMPARE
$920.87CAD / Each
COMPARE
$889.93CAD / Each