This product is now discontinued, View Related Products
Cole-Parmer Low-Cost Ultrasonic Cleaner With Timer
Economical compact cleaner is perfect for cleaning small components
This 15-oz ultrasonic cleaner measures only 8 ¾''L x 4 ½''W (22.2 x 11.4 cm) — ideal for small benchtop areas. The stainless steel tank is sealed inside an impact resistant plastic housing to prevent leaks. The cleaner features a five-minute timer that shuts off automatically. Operates at a cleaning frequency of 40 kHz.
$315.31 - $350.67CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Power (VAC)
Availability Pricing
COMPARE
0.125 117
$315.31CAD / Each
COMPARE
0.125 220
$350.67CAD / Each