This product is now discontinued, View Related Products
Cole-Parmer Liquid Chromatography Detectors
Sensitive, affordable detectors
  • Detectors for HPLC and low-pressure chromatography
Replace your failed component or augment your liquid chromatography system with these precision UV/Vis detectors. All wetted parts are PEEK, PTFE, or Tefzel™ for excellent chemical resistance and biocompatibility. Integrate detector into your data stream via analog output.
Variable Wavelength DetectorsThese versatile instruments have double beam optics, chemical-resistant fluid contact surfaces, and a small footprint. Select your UV/Vis wavelength (195 to 800 nm) using front panel scrolling or remote computer-driven contact closures. Wavelength accuracy: 2 nm, reproducibility: 1 nm. Digital readout and analog outputs for wavelength and absorbance
$825.10 - $8,999.47CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Wavelength (nm) Min Wavelength (nm) Power (VAC)
Availability Pricing
COMPARE
$825.10CAD / Each
COMPARE
254 254 115
$8,999.47CAD / Each
COMPARE
254 254 230
$8,999.47CAD / Each