This product is now discontinued, View Related Products
Cole-Parmer LED Fiber Optic Illuminators 60653
LED bulb provides energy efficiency with high light output
  • Small footprint easily fits on your benchtop
Designed for general microscopy, assembly illumination, machine vision, and boroscope applications. High-power white LEDs provide consistent color temperature and higher light output and longer life than conventional 150-watt fiber optic illuminator bulbs. Adjust the light output with the 15-step intensity control.
$2,012.65 - $2,590.22CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Power Cord Included Description
Availability Pricing
COMPARE
115 VAC, US plug LED Fiber Optic Illuminator; 115 VAC N. American Power Cord
$2,012.65CAD / Each
COMPARE
230 VAC, European plug LED Fiber Optic Illuminator; 230 VAC European Power Cord
$2,012.65CAD / Each
COMPARE
115 VAC, US plug LED Fiber Optic Illuminator w/Gooseneck; 115 VAC Cord
$2,266.07CAD / Each
COMPARE
230 VAC, European plug LED Fiber Optic Illuminator w/Gooseneck; 230 VAC Cord
$2,299.96CAD / Each
COMPARE
115 VAC, US plug LED Fiber Optic Illuminator w/Dual Gooseneck; 115 VAC Cord
$2,417.10CAD / Each
COMPARE
230 VAC, European plug LED Fiber Optic Illuminator w/Dual Gooseneck; 230 VAC Cord
$2,472.35CAD / Each
COMPARE
115 VAC, US plug LED Fiber Optic Illuminator w/60-mm Ring Light; 115 VAC Cord
$2,589.48CAD / Each
COMPARE
230 VAC, European plug LED Fiber Optic Illuminator w/60-mm Ring Light; 230 VAC Cord
$2,589.48CAD / Each
COMPARE
115 VAC, US plug LED Fiber Optic Illuminator w/66-mm Ring Light; 115 VAC Cord
$2,590.22CAD / Each
COMPARE
230 VAC, European plug LED Fiber Optic Illuminator w/66-mm Ring Light; 230 VAC Cord
$2,590.22CAD / Each
Showing 1- 10 of 10