Cole-Parmer® HO-400 Series Laboratory Homogenizer
Process virtually any laboratory sample in seconds
  • Variable-speed motor performs from 5,000 to 25,000 rpm
  • Processing range from 200 uL to 5 L
  • Digital, programmable speed and time controls
The Cole-Parmer homogenizers are well-suited for a wide range of processing applications - use as a handheld unit or post-mount to the optional stand for hands-free operation. Use these homogenizers with either stainless steel rotor-stator generators or disposable plastic probes.
Order components and lab stand (04727-89) separately.
$3,757.14CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Sample Size (mL) Power (VAC)
Availability Pricing
COMPARE
25000 0.2 110
$3,757.14CAD / Each
COMPARE
25000 0.2 220
$3,757.14CAD / Each