Cole-Parmer® HO-200 Series Tissue Homogenizers
Lightweight and powerful motor ideal for small samples
  • Variable-speed motor performs up to 30,000 rpm
  • Ergonomically designed to reduce hand strain
The Cole-Parmer HO-200 series homogenizers are well-suited for a wide range of processing applications —use as a handheld unit or post-mount to the optional stand for hands-free operation. Use these homogenizers with either the stainless steel rotor-stator generators or the plastic tip probes (tip adapter 04727-88 required). Order components separately.
These homogenizers process a wide range of samples from 0.05 mL to 100 mL. They are ergonomically designed to reduce hand strain during extended periods of handheld operation.
$1,617.78 - $1,809.32CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Sample Size (mL) Power (VAC) Description
Availability Pricing
COMPARE
100 115 Tissue Homogenizer with US Plug; 115V
$1,809.32CAD / Each
COMPARE
100 220 Tissue Homogenizer with EU Plug; 220V
$1,809.32CAD / Each
COMPARE
100 220 Tissue Homogenizer with UK plug
$1,617.78CAD / Each