Cole-Parmer® IPSW-400 Series Washdown Industrial Bench Scales
Withstands splashes, spills, and sprays
  • IP66-rated housing and indicator are suitable for washdown environments
  • Rugged stainless-steel construction and sealed keypad resist damage from dirt and moisture
  • Long 70-hour battery life means fewer recharging cycles
  • Capacity tracker prevents overload conditions
  • Selectable sample size makes repetitive tasks more efficient
Scales feature weighing, parts counting, percentage, checkweighing, dynamic, and accumulation functions.
$1,508.01 - $2,867.22CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
COMPARE
15 30 115 / 220
$1,844.68CAD / Each
COMPARE
50 100 115 / 220
$2,039.17CAD / Each
COMPARE
75 150 115 / 220
$2,459.09CAD / Each
COMPARE
150 300 115 / 220
$2,867.22CAD / Each
COMPARE
15 30 220
$1,508.01CAD / Each
COMPARE
50 100 220
$1,667.14CAD / Each
COMPARE
75 150 220
$2,011.18CAD / Each
COMPARE
150 300 220
$2,345.64CAD / Each
COMPARE
25 50 115 / 220
$2,459.09CAD / Each
COMPARE
25 50 220
$2,011.18CAD / Each
Showing 1- 10 of 10