This product is now discontinued, View Related Products
Cole-Parmer Horizontal Mini-Gel Electrophoresis Systems
Compact, leak-free casting—ideal for quick screening
Gel beds and trays are UV-transparent acrylic—scan gels at 275 to 325 nm without removing them from the unit. The tapered baffle design eliminates the need for sealing or taping. Compact format saves time and costly reagents. Optional gel trays and casters let you pour six gels simultaneously. Order combs, gel trays, and gel casters separately.
$519.66 - $588.47CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Description
Availability Pricing
COMPARE
150 Horizontal Mini-gel unit, 5.5 x 8.5 cm gel bed
$519.66CAD / Each
COMPARE
250 Horizontal Mini-gel unit, 7.5 x 10 cm gel bed
$588.47CAD / Each