This product is now discontinued, View Related Products
Cole-Parmer Horizontal Mid-Size Gel Systems
Reduced gel path length for rapid sequencing of samples
Tapered-baffle design eliminates the need for sealing or taping. UV-transparent acrylic gel tray has ''anchors'' to prevent gel from floating during electrophoresis. Optional gel casters let you pour gels outside of the unit, and to pour two gels simultaneously.
$742.22 - $1,088.10CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Description
Availability Pricing
COMPARE
$1,088.10CAD / Each
COMPARE
700 Midi-horizontal gel units, 14 x 10 cm gel bed
$742.22CAD / Each
COMPARE
750 Midi-horizontal gel units, 14 x 16 cm gel bed
$1,039.11CAD / Each