This product is now discontinued, View Related Products
Cole-Parmer Horizontal Mid-Size Gel Casters
Gel Casters allow you to precast consistently flat, even gels using the gel trays sold seperately. Casters accommodate either one or two trays. Casters feature a three-point leveling base and soft, coated neoprene side gaskets that secure trays in place and effectively seal their ends during casting — eliminating the need for taping.
$81.04 - $344.77CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
$81.04CAD / Each
COMPARE
$344.77CAD / Each
COMPARE
Mid-Size System 1-Place Gel Caster Tray for 28560-00
$129.66CAD / Each
COMPARE
Mid-Size System 2-Place Gel Caster Tray for 28560-00
$223.96CAD / Each
COMPARE
Mid-Size System 1-Place Gel Caster Tray for 28560-10
$129.66CAD / Each
COMPARE
Mid-Size System 2-Place Gel Caster Tray for 28560-10
$216.59CAD / Each
COMPARE
Submarine Gel System 1-Place Gel Caster Tray for 28561-00
$129.66CAD / Each
COMPARE
Submarine Gel System 2-Place Gel Caster Tray for 28561-00
$237.22CAD / Each
Showing 1- 8 of 8