Cole-Parmer® Griffin Graduated Polypropylene Beakers
No more difficult readings
  • Easy-to-read blue graduations
  • Autoclavable
Polypropylene (PP) beakers are ideal for general laboratory use. Screened-on blue graduations are easy to read and give approximate graduations. Offer superior chemical resistance and withstand temperatures to 275°F (135°C).
$68.51 - $106.82CAD / Case of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
50 10 Polypropylene(PP)
$80.30CAD / Case of 12
COMPARE
100 20 Polypropylene(PP)
$80.30CAD / Case of 12
COMPARE
250 50 Polypropylene(PP)
$68.51CAD / Case of 6
COMPARE
400 50 Polypropylene(PP)
$106.82CAD / Case of 6
COMPARE
600 100 Polypropylene(PP)
$77.35CAD / Case of 4
COMPARE
1000 100 Polypropylene(PP)
$75.88CAD / Case of 3