Cole-Parmer® Essentials Graduated Cylinders, Polypropylene
Don’t guess at volume
  • Read with ease—excellent transparency and raised graduations Durable, one-piece construction Maintain cleanliness—autoclavable
These polypropylene (PP) cylinders feature excellent transparency. Molded in one piece for enhanced durability. Cylinders have raised graduations for easy reading. Autoclavable.
$22.84 - $175.33CAD / Pkg of 2
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
10 0.2 Polypropylene
$22.84CAD / Pkg of 2
COMPARE
25 0.5 Polypropylene
$25.78CAD / Pkg of 2
COMPARE
50 1 Polypropylene
$23.57CAD / Pkg of 2
COMPARE
100 1 Polypropylene
$33.15CAD / Pkg of 2
COMPARE
250 2 Polypropylene
$24.31CAD / Pkg of 1
COMPARE
500 5 Polypropylene
$31.68CAD / Pkg of 1
COMPARE
1000 10 Polypropylene
$33.15CAD / Pkg of 1
COMPARE
2000 20 Polypropylene
$175.33CAD / Pkg of 2
Showing 1- 8 of 8