Environmental Express General-Purpose Glass Body Combination pH/ORP Electrodes

Faster response and increased stability across a pH range of 0 to 14
  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
$232.06 - $535.28CAD / Each

13 variations of this product are available.

See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Applications Electrode Type Description
Availability Pricing
COMPARE
High temp & Chemicals Single junction pH electrode, Refillable/SJ/Glass/Extra-Long/Thin/326x8mm; BNC
$386.40CAD / Each
COMPARE
General purpose Single junction ORP Electrode, Sealed, Waste water, reaction monitoring
$320.24CAD / Each
COMPARE
Dirty Samples, Waste water Double junction ORP Electrode, Sealed, Waste water, reaction monitoring
$431.26CAD / Each
COMPARE
General purpose-lab Single junction General purpose-lab, pH electrode, Refillable
$326.87CAD / Each
COMPARE
Hi temp and pH with high Sodium Single junction pH Probe, Refillable, High-temp, pH with high Na+
$345.44CAD / Each
COMPARE
General purpose-Thin neck Single junction pH electrode, Refillable, long and thin body
$288.56CAD / Each
COMPARE
Flat surface or low volume sampling Double junction pH electrode, Refillable, flat-surface
$535.28CAD / Each
COMPARE
Larger test tubes Single junction pH Probe, Refillable, long and thin body
$338.81CAD / Each
COMPARE
Small test tubes sampling Single junction pH electrode, Refillable, long and thin body
$361.28CAD / Each
COMPARE
Rugged lab and field Single junction pH electrode, Refillable low maintenance
$232.06CAD / Each
COMPARE
Rugged lab and field Single junction pH electrode, general purpose, combination, glass body, Delrin shield, BNC
$292.03CAD / Each
COMPARE
Research, TRIS, sulfates, proteins Double junction Combination pH electrode, Glass, Refillable, Double-Junction, BNC
$383.30CAD / Each
COMPARE
Rugged for TRIS, sulfates, proteins Double junction Combination pH Electrode, Glass, Double-Junction, BNC
$388.75CAD / Each
Showing 1- 13 of 13

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat