This product is now discontinued, View Related Products
Polyscience Flow-Through Refrigerated Chillers
Designed to run at maximum cooling rate
  • High performance auxiliary cooling systems
Use these flow-through refrigerated chillers with circulators, tanks, and open baths that either lack refrigeration or need additional cooling. You can use these units in circulating systems where a pressure and/or suction pump is supplied. We recommend flow rates of 3 to 25 liters/minute at a maximum of 12 psi. These chillers are designed to run at maximum cooling rate. Temperature control is provided by an external circulator—order separately.
Operate using a fluid such as Dynalene bath fluid or an ethylene glycol/water mixture.
$2,283.75 - $3,867.65CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
Flow-through Refrgerated Chillers; range, -20 to 40°C; 120V; 60Hz
$2,283.75CAD / Each
COMPARE
Flow-through Refrgerated Chillers; range, -20 to 40°C; 240V; 50Hz
$2,283.75CAD / Each
COMPARE
40 -25 Flow-Through Chiller, -25 to 40 C, 120 VAC/60 Hz
$3,867.65CAD / Each
COMPARE
40 -25 Flow-Through Chiller, -25 to 40 C, 240 VAC/50 Hz
$3,867.65CAD / Each