Cole-Parmer® Flow-Through Cells
Ideal for fermentation applications
  • Accurate to ±0.1 pH units
  • Choose 316 stainless steel cell for long-lasting durability
Measure the pH of high-purity water without the problems usually associated with low-ionic strength solutions.
$277.00 - $282.89CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH flow-through cell, 6000 uL, 7 mm ID, glass
$277.00CAD / Each
COMPARE
pH flow-through cell, 800 uL, 3 mm ID, glass
$282.89CAD / Each