This product is now discontinued, View Related Products
Cole-Parmer Filter Pipette Tips 877
These state-of-the-art polypropylene pipette tips feature a unique, hydrophobic super filter. The inert filter prevents liquid bypass in the event of over-pipetting and doesn't seal or trap liquid—allowing for non-contaminated sample recovery. Filters also eliminate trace carryover and contamination by aerosols generated during sample aspiration.
Autoclavable tips are available either non-sterile or gamma-irradiation sterilized, and packaged in racks of 96 or 100. Each rack is stackable, shrink-wrapped with a tear-off seal, and labeled with pipette type and lot number.
$54.15 - $209.22CAD / Pkg
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL)
Availability Pricing
COMPARE
10
$110.50CAD / Pkg
COMPARE
10
$207.01CAD / Pkg
COMPARE
10
$106.08CAD / Pkg
COMPARE
10
$207.01CAD / Pkg
COMPARE
$138.50CAD / Pkg
COMPARE
30
$207.01CAD / Pkg of 960
COMPARE
$54.15CAD / Pkg
COMPARE
$116.40CAD / Pkg
COMPARE
100
$110.50CAD / Pkg
COMPARE
100
$206.27CAD / Pkg of 10
COMPARE
$138.50CAD / Pkg
COMPARE
200
$207.01CAD / Pkg of 960
COMPARE
250
$57.78CAD / Pkg
COMPARE
$61.81CAD / Pkg
COMPARE
1000
$101.66CAD / Pkg
COMPARE
1000
$203.33CAD / Pkg
COMPARE
1000
$114.92CAD / Pkg
COMPARE
$120.82CAD / Pkg
COMPARE
10
$165.02CAD / Pkg
COMPARE
200
$170.91CAD / Pkg
Showing 1 - 20 of 21