Foxx Life Sciences Filling and Venting Carboy Caps
Eliminate pouring expensive solutions with these aseptic caps
  • Retrofit to your system—choose from common carboy cap sizes
  • Designed to accept connectors and air vent filters
  • Polypropylene and high-purity silicone materials may be autoclaved
Foxx Life Sciences™ VersaCap® Filling and Venting closures are aseptic handling solutions for pharmaceutical, biotechnology, and laboratory liquid transfer applications. Eliminate pouring of high valued products through this leak proof aseptic system.
$169.81CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Tubing ID
Availability Pricing
Foxx Life Sciences 205-6103-CP Carboy Cap, 83 mm, 2 x 1/4" Hose Barb, with Tubing RK-06059-33 Mfr # 205-6103-RLS
COMPARE
83B 2 x 1/4\" ID
$169.81CAD / Each