This product is now discontinued, View Related Products
Cole-Parmer Field Benchtop Turbidity Meters
Front-panel dials simplify your turbidity measurements
  • Additional sample cells within easy reach — stored directly in the case with the field meters
Use these meters to verify water cleanliness in public water systems and to control chemicals through water treatment. Other applications include chemically analyzing chlorides and other elements through controlled precipitation, checking filter efficiency, and isolating filter backwash problems. Meters measure particulate content of water in three ranges. All results are displayed in NTU on the 1/2"H (1.3 cm) LCD.
$2,216.72 - $2,226.29CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Turbidity Resolution
Availability Pricing
COMPARE
0 to 199.9 0.1 NTU
$2,226.29CAD / Each
COMPARE
0 to 199.9 0.1 NTU
$2,216.72CAD / Each