This product is now discontinued, View Related Products
Cole-Parmer Exhaust Filters for Solvent Waste Systems
Affordable active carbon filter solution for HPLC liquid waste disposal
Use this activated carbon exhaust filter with your existing HPLC solvent waste systems to trap volatile organic compounds from being released into the air. This protects workers from inhaling dangerous chemicals and provides an easy mechanism of disposal for vapor wastes.
$128.92 - $1,019.59CAD / Pkg of 2
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Filter Diameter (in) Filter Length (in) Number of Cartridges
Availability Pricing
COMPARE
1.889 6.97 2
$226.90CAD / Pkg of 2
COMPARE
1.889 6.97 1
$128.92CAD / Each
COMPARE
1.889 6.97 5
$380.13CAD / Pkg of 5
COMPARE
2.677 10.5 1
$400.76CAD / Each
COMPARE
2.724 14 1
$607.04CAD / Each
COMPARE
2.677 10.5 2
$670.39CAD / Pkg of 2
COMPARE
2.724 14 2
$1,019.59CAD / Pkg of 2
Showing 1- 7 of 7