Cole-Parmer® Essentials Sterile Lab Sampling Scoops with Lids, Polystyrene
Lid keeps sample contained within sampling scoop
  • Gamma irradiation sterilized SAL 10-6
  • Meets FDA guidelines for food, drug, and cosmetic sampling
  • Flat bottom for use as weigh boat
  • Individually sealed easy-to-open bags
A transparent lid snaps securely onto these sterile, disposable lab scoops to keep the sample contained while allowing it to be easily viewed. No additional sample bag or container is needed. Manufactured from sturdy FDA-grade polystyrene plastic, these sampling scoops have an ergonomic, comfortably curved handle to make sample retrieval easy. The lab scoops have a flat bottom to allow them to be placed on a surface without spillage and to be used as a weighing dish.
These individually bagged laboratory scoops are gamma irradiation sterilized SAL 10-6; are free of BSE/TSE, heavy metals, BPA, and phthalates; and lot traced. A Certificate of Irradiation is available upon request. Specified volumes are approximate and not intended for measuring specific volumes.
$169.44 - $184.17CAD / Case of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
60 20.21 3.92
$169.44CAD / Case of 50
COMPARE
120 22.01 4.91
$184.17CAD / Case of 50